COM448686663 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-chlorophenyl)propanoate

  • MDL number MFCD27578435
  • Cas Number : 851967-77-6
  • Molecular Weight : 313.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1595

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1595
Availability details for
Quantity
  • Availability
  • Add to Cart