COM448686665 Combi-Blocks, Inc.

methyl 2-(2-{4-[(R)-(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetate dihydrochloride

  • MDL number MFCD27578437
  • Cas Number : 1820572-16-4
  • Molecular Weight : 475.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1597

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1597
Availability details for
Quantity
  • Availability
  • Add to Cart