COM448686668 Combi-Blocks, Inc.

methyl 7-fluoro-8-(4-hydroxypiperidin-1-yl)-12-methyl-4-oxo-1-azatricyclo[7.3.1.0^{5,13}]trideca-2,5(13),6,8-tetraene-3-carboxylate

  • MDL number MFCD27578439
  • Cas Number : 129672-74-8
  • Molecular Weight : 374.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1601
Availability details for
Quantity
  • Availability
  • Add to Cart