COM448686669 Combi-Blocks, Inc.

Methyl (R)-2-Azetidinecarboxylate

  • MDL number MFCD22397976
  • Cas Number : 43041-13-0
  • Molecular Weight : 151.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1602

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1602
Availability details for
Quantity
  • Availability
  • Add to Cart