COM448686671 Combi-Blocks, Inc.

methyl 1-[(4-methylbenzene)sulfonyl]imidazole-4-carboxylate

  • MDL number MFCD27578440
  • Cas Number : 1820716-97-9
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1604
Availability details for
Quantity
  • Availability
  • Add to Cart