COM448686675 Combi-Blocks, Inc.

Methyl 4-fmoc-piperazine-2-carboxylate

  • MDL number MFCD01632467
  • Cas Number : 647004-81-7
  • Molecular Weight : 366.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1608

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1608
Availability details for
Quantity
  • Availability
  • Add to Cart