COM448686679 Combi-Blocks, Inc.

methyl 5,7-dichloro-4-hydroxyquinoline-3-carboxylate

  • MDL number MFCD27578445
  • Cas Number : 1820685-13-9
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1614

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1614
Availability details for
Quantity
  • Availability
  • Add to Cart