COM448686684 Combi-Blocks, Inc.

2-Benzoylamino-benzoic acid methyl ester

  • MDL number MFCD00089143
  • Cas Number : 7510-49-8
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1622

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1622
Availability details for
Quantity
  • Availability
  • Add to Cart