COM448686692 Combi-Blocks, Inc.

methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-cyclopropylacetate

  • MDL number MFCD27578452
  • Cas Number : 936097-43-7
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1631

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1631
Availability details for
Quantity
  • Availability
  • Add to Cart