COM448686696 Combi-Blocks, Inc.

Methyl 2-amino-2-methylbutanoate

  • MDL number MFCD12151626
  • Cas Number : 87162-70-7
  • Molecular Weight : 131.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1638
Availability details for
Quantity
  • Availability
  • Add to Cart