COM448686697 Combi-Blocks, Inc.

4-Hydroxy-7-methoxyquinoline-3-carboxylic acid methyl ester

  • MDL number MFCD22418577
  • Cas Number : 1448260-35-2
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1640
Availability details for
Quantity
  • Availability
  • Add to Cart