COM448686699 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-cyclopentylacetate

  • MDL number MFCD27578456
  • Cas Number : 196206-09-4
  • Molecular Weight : 257.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1644

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1644
Availability details for
Quantity
  • Availability
  • Add to Cart