COM448686700 Combi-Blocks, Inc.

methyl (1S)-1,2,3,4-tetrahydronaphthalene-1-carboxylate

  • MDL number MFCD27578457
  • Cas Number : 900789-32-4
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1646
Availability details for
Quantity
  • Availability
  • Add to Cart