COM448686713 Combi-Blocks, Inc.

Methyl 1-phenyl-1h-imidazole-4-carboxylate

  • MDL number MFCD20921991
  • Cas Number : 116343-89-6
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1666

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1666
Availability details for
Quantity
  • Availability
  • Add to Cart