COM448686717 Combi-Blocks, Inc.

3-Methoxy-5-(trifluoromethoxy)benzoic acid methyl ester

  • MDL number MFCD18399560
  • Cas Number : 1261583-60-1
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1671
Availability details for
Quantity
  • Availability
  • Add to Cart