COM448686725 Combi-Blocks, Inc.

Namiki bbv-27181239

  • MDL number MFCD12127025
  • Cas Number : 66328-82-3
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1681
Availability details for
Quantity
  • Availability
  • Add to Cart