COM448686726 Combi-Blocks, Inc.

2-(2,4-Difluorophenoxy)-acetic acid methyl ester

  • MDL number MFCD10694494
  • Cas Number : 449811-63-6
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1684

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1684
Availability details for
Quantity
  • Availability
  • Add to Cart