COM448686728 Combi-Blocks, Inc.

methyl 2-fluoro-4-phenylbenzoate

  • MDL number MFCD27578462
  • Cas Number : 505083-19-2
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1687
Availability details for
Quantity
  • Availability
  • Add to Cart