COM448686731 Combi-Blocks, Inc.

Methyl 2-amino-3-(4-hydroxy-3-methoxyphenyl)propanoate

  • MDL number MFCD19661232
  • Cas Number : 113948-28-0
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1690

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1690
Availability details for
Quantity
  • Availability
  • Add to Cart