COM448686732 Combi-Blocks, Inc.

4-tert-Butyl-2-ethoxy-benzoic acid methyl ester

  • MDL number MFCD11501371
  • Cas Number : 870007-39-9
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1691

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1691
Availability details for
Quantity
  • Availability
  • Add to Cart