COM448686733 Combi-Blocks, Inc.

Methyl (2s)-2-amino-3-naphthylpropanoate

  • MDL number MFCD08058289
  • Cas Number : 119357-91-4
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1692

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1692
Availability details for
Quantity
  • Availability
  • Add to Cart