COM448686734 Combi-Blocks, Inc.

Methyl (2S)-2-amino-3-(2-nitrophenyl)propanoate

  • MDL number MFCD09255793
  • Cas Number : 1212908-89-8
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1693
Availability details for
Quantity
  • Availability
  • Add to Cart