COM448686736 Combi-Blocks, Inc.

methyl 2-(5-bromo-2-oxo-3,4-dihydro-1-benzopyran-4-yl)acetate

  • MDL number MFCD27578463
  • Cas Number : 1820711-99-6
  • Molecular Weight : 299.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1696

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1696
Availability details for
Quantity
  • Availability
  • Add to Cart