COM448686738 Combi-Blocks, Inc.

Methyl (2s)-2-amino-2-(thiophen-2-yl)acetate

  • MDL number MFCD20719696
  • Cas Number : 53585-44-7
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1698
Availability details for
Quantity
  • Availability
  • Add to Cart