COM448686740 Combi-Blocks, Inc.

methyl 1-{[(tert-butoxy)carbonyl]amino}-3,6,9,12-tetraoxapentadecan-15-oate

  • MDL number MFCD27578465
  • Cas Number : 1820674-09-6
  • Molecular Weight : 379.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1700
Availability details for
Quantity
  • Availability
  • Add to Cart