COM448686741 Combi-Blocks, Inc.

4-Fluoro-4'-(trifluoromethoxy)biphenyl-3-carboxylic acid methyl ester

  • MDL number MFCD18408101
  • Cas Number : 1261744-05-1
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1701
Availability details for
Quantity
  • Availability
  • Add to Cart