COM448686745 Combi-Blocks, Inc.

1-tert-butyl 4-methyl (2S)-2-aminobutanedioate

  • MDL number MFCD02094358
  • Cas Number : 104072-49-3
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1709

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1709
Availability details for
Quantity
  • Availability
  • Add to Cart