COM448686746 Combi-Blocks, Inc.

Methyl 3-methyl-5-(trifluoromethyl)benzoate

  • MDL number MFCD14698072
  • Cas Number : 161622-18-0
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1710

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1710
Availability details for
Quantity
  • Availability
  • Add to Cart