COM448686749 Combi-Blocks, Inc.

methyl (2S)-2-amino-3-{[(4-methoxyphenyl)methyl]sulfanyl}-3-methylbutanoate

  • MDL number MFCD27578468
  • Cas Number : 1820576-01-9
  • Molecular Weight : 283.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1715

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1715
Availability details for
Quantity
  • Availability
  • Add to Cart