COM448686751 Combi-Blocks, Inc.

methyl 4,5-dimethoxy-2-(2-oxopyrrolidin-1-yl)benzoate

  • MDL number MFCD27578470
  • Cas Number : 1820683-98-4
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1722

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1722
Availability details for
Quantity
  • Availability
  • Add to Cart