COM448686758 Combi-Blocks, Inc.

methyl 4-tert-butyl-3-nitrobenzoate

  • MDL number MFCD27578472
  • Cas Number : 91641-96-2
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1731
Availability details for
Quantity
  • Availability
  • Add to Cart