COM448686759 Combi-Blocks, Inc.

methyl (2E)-3-[5-bromo-2-(trifluoromethoxy)phenyl]prop-2-enoate

  • MDL number MFCD27578473
  • Cas Number : 1820748-61-5
  • Molecular Weight : 325.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1732
Availability details for
Quantity
  • Availability
  • Add to Cart