COM448686761 Combi-Blocks, Inc.

methyl 3-(4-ethylphenyl)benzoate

  • MDL number MFCD05980136
  • Cas Number : 1049118-09-3
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1735
Availability details for
Quantity
  • Availability
  • Add to Cart