COM448686762 Combi-Blocks, Inc.

Methyl 3'-methoxy[1,1'-biphenyl]-3-carboxylate

  • MDL number MFCD06204192
  • Cas Number : 128460-75-3
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1739
Availability details for
Quantity
  • Availability
  • Add to Cart