COM448686763 Combi-Blocks, Inc.

methyl 2-(4-bromo-2-fluorophenyl)-2-methylpropanoate

  • MDL number MFCD27578475
  • Cas Number : 749929-35-9
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1740
Availability details for
Quantity
  • Availability
  • Add to Cart