COM448686764 Combi-Blocks, Inc.

1-tert-butyl 3-methyl (3r,4s)-4-(4-fluorophenyl)pyrrolidine-1,3-dicarboxylate, rac, trans

  • MDL number MFCD27578476
  • Cas Number : 1820574-66-0
  • Molecular Weight : 323.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1741
Availability details for
Quantity
  • Availability
  • Add to Cart