COM448686772 Combi-Blocks, Inc.

Methyl (2s)-2-amino-3-(furan-2-yl)propanoate

  • MDL number MFCD10565764
  • Cas Number : 146725-85-1
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1753
Availability details for
Quantity
  • Availability
  • Add to Cart