COM448686773 Combi-Blocks, Inc.

methyl 2-chloro-5-(2-formylphenyl)benzoate

  • MDL number MFCD27578480
  • Cas Number : 1820673-88-8
  • Molecular Weight : 274.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1754
Availability details for
Quantity
  • Availability
  • Add to Cart