COM448686774 Combi-Blocks, Inc.

Methyl 2-(1h-imidazol-1-yl)benzoate

  • MDL number MFCD11870283
  • Cas Number : 246046-82-2
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1755
Availability details for
Quantity
  • Availability
  • Add to Cart