COM448686778 Combi-Blocks, Inc.

(S)-Dimethyl 2-(tert-butoxycarbonylamino)succinate

  • MDL number MFCD04116316
  • Cas Number : 55747-84-7
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1761

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1761
Availability details for
Quantity
  • Availability
  • Add to Cart