COM448686779 Combi-Blocks, Inc.

methyl (2S,3R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}butanoate

  • MDL number MFCD27578483
  • Cas Number : 137719-70-1
  • Molecular Weight : 323.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1763

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1763
Availability details for
Quantity
  • Availability
  • Add to Cart