COM448686780 Combi-Blocks, Inc.

triethylamine methyl (2S)-3-hydroxy-2-[(triphenylmethyl)amino]propanoate

  • MDL number MFCD27578484
  • Cas Number : 1820570-35-1
  • Molecular Weight : 462.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1764
Availability details for
Quantity
  • Availability
  • Add to Cart