COM448686786 Combi-Blocks, Inc.

methyl 2-chloro-4-(3-methylphenyl)benzoate

  • MDL number MFCD27578486
  • Cas Number : 1820648-48-3
  • Molecular Weight : 260.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1772
Availability details for
Quantity
  • Availability
  • Add to Cart