COM448686787 Combi-Blocks, Inc.

methyl 4-(3,5-dimethoxyphenyl)benzoate

  • MDL number MFCD27578487
  • Cas Number : 913647-92-4
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1774

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1774
Availability details for
Quantity
  • Availability
  • Add to Cart