COM448686788 Combi-Blocks, Inc.

methyl 3-(3-methylphenyl)-5-(trifluoromethyl)benzoate

  • MDL number MFCD27578488
  • Cas Number : 1820666-87-2
  • Molecular Weight : 294.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1775
Availability details for
Quantity
  • Availability
  • Add to Cart