COM448686789 Combi-Blocks, Inc.

methyl 5-(4-nitrophenyl)-1,3-oxazole-4-carboxylate

  • MDL number MFCD27578490
  • Cas Number : 72030-86-5
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1777
Availability details for
Quantity
  • Availability
  • Add to Cart