COM448686798 Combi-Blocks, Inc.

methyl (2R)-2-amino-3-[1-(triphenylmethyl)imidazol-4-yl]propanoate

  • MDL number MFCD27578495
  • Cas Number : 164166-96-5
  • Molecular Weight : 411.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1791
Availability details for
Quantity
  • Availability
  • Add to Cart