COM448686800 Combi-Blocks, Inc.

Methyl 2-(benzyloxy)-5-bromobenzoate

  • MDL number MFCD11112970
  • Cas Number : 860000-78-8
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1794

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1794
Availability details for
Quantity
  • Availability
  • Add to Cart