COM448686801 Combi-Blocks, Inc.

methyl (3R)-3-aminohex-5-enoate hydrochloride

  • MDL number MFCD27578497
  • Cas Number : 1820571-33-2
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1796
Availability details for
Quantity
  • Availability
  • Add to Cart