COM448686802 Combi-Blocks, Inc.

1-(3-Chloro-4-methyl-phenyl)-5-oxo-pyrrolidine-3-carboxylic acid methyl ester

  • MDL number MFCD01441280
  • Cas Number : 345367-42-2
  • Molecular Weight : 267.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1797
Availability details for
Quantity
  • Availability
  • Add to Cart